Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004079.4 | NP_004070.3 | 4 | 642 | ||
GenBank mRNA | AK226095.1 | - | 4 | 504 | ||
AK314482.1 | - | 4 | 483 | |||
BC002642.2 | - | 4 | 443 | 3610589 | ||
CR541676.1 | - | 3 | 381 | |||
M86553.1 | - | 3 | 387 | |||
M90696.1 | - | 4 | 517 | |||
S93414.1 | - | 3 | 387 |