Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173806.3 | NP_776167.2 | 3 | 470 | ||
NR_033694.1 | 5 | 721 | ||||
XM_006721029.3 | XP_006721092.1 | 4 | 674 | |||
XM_011545785.2 | XP_011544087.1 | 4 | 677 | |||
XM_017023109.1 | XP_016878598.1 | 3 | 496 | |||
XM_017023110.1 | XP_016878599.1 | 5 | 735 | |||
GenBank mRNA | BC021724.1 | - | 5 | 726 | 4829370 | |
BC028605.1 | - | 5 | 726 | 5265642 | ||
BC039562.1 | - | 5 | 718 | 5169242 |