Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPY19L2P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003551.1 | 3 | 1676 | |||
GenBank mRNA | BC031277.1 | - | 3 | 1633 | ||
BC047471.1 | - | 3 | 1624 | |||
BX537815.1 | - | 3 | 1676 | |||
DQ287934.1 | - | 3 | 1676 |
Gene Symbol: DPY19L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173812.4 | NP_776173.3 | 22 | 3713 | ||
XM_006719352.2 | XP_006719415.1 | 22 | 3639 | |||
XM_011538215.2 | XP_011536517.1 | 20 | 3179 | |||
XM_011538218.2 | XP_011536520.1 | 15 | 2934 | |||
XM_017019188.1 | XP_016874677.1 | 22 | 3760 | |||
XM_017019189.1 | XP_016874678.1 | 21 | 3700 | |||
XM_017019190.1 | XP_016874679.1 | 21 | 3634 | |||
XM_017019191.1 | XP_016874680.1 | 20 | 3610 | |||
XM_017019192.1 | XP_016874681.1 | 20 | 3598 | |||
XM_017019193.1 | XP_016874682.1 | 21 | 3295 | |||
XM_017019194.1 | XP_016874683.1 | 21 | 3322 | |||
XM_017019195.1 | XP_016874684.1 | 22 | 3370 | |||
XM_017019196.1 | XP_016874685.1 | 20 | 3459 | |||
XM_017019197.1 | XP_016874686.1 | 21 | 3593 | |||
XM_017019198.1 | XP_016874687.1 | 18 | 3314 | |||
XM_017019199.1 | XP_016874688.1 | 18 | 3383 | |||
XM_017019201.1 | XP_016874690.1 | 21 | 3556 | |||
XM_017019202.1 | XP_016874691.1 | 20 | 3169 | |||
XM_017019203.1 | XP_016874692.1 | 20 | 3391 | |||
XM_017019204.1 | XP_016874693.1 | 21 | 3629 | |||
XM_017019205.1 | XP_016874694.1 | 18 | 3483 | |||
GenBank mRNA | AL833344.1 | - | 22 | 3640 | ||
AY358792.1 | - | 22 | 3631 | |||
DB514316.1 | - | 1 | 70 |
Gene Symbol: DPY19L2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648666.1 | - | 18 | 3008 |