Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HECTD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109662.3 | NP_001103132.3 | 25 | 3930 | ||
GenBank mRNA | AB777655.1 | - | 25 | 4252 | ||
AK091473.1 | - | 15 | 2093 | |||
BC101482.1 | - | 14 | 2009 | |||
BC143384.1 | - | 14 | 2003 | |||
BC143388.1 | - | 14 | 2036 |