Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267774.1 | NP_001254703.1 | 5, 6 | 706 | ||
NM_001267775.1 | NP_001254704.1 | 5, 6 | 505 | |||
NM_001267776.1 | NP_001254705.1 | 4, 5 | 469 | |||
NM_174887.3 | NP_777547.1 | 5, 6 | 587 | |||
GenBank mRNA | AY224601.1 | - | 5, 6 | 655 | ||
BC002640.2 | - | 5, 6 | 567 | 3609552 | ||
BC038094.1 | - | 4, 5 | 435 | 3907361 | ||
BE619475.1 | - | 5, 6 | 441 |