Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135058.1 | NP_001128530.1 | 3 | 537 | ||
NM_004086.2 | NP_004077.1 | 4 | 213 | |||
XM_017021071.1 | XP_016876560.1 | 4 | 447 | |||
XM_017021072.1 | XP_016876561.1 | 3 | 586 | |||
GenBank mRNA | AF006740.1 | - | 4 | 189 | ||
AK123362.1 | - | 2 | 343 | |||
AK292724.1 | - | 4 | 189 | |||
AW629100.1 | - | 3 | 537 | |||
AY358900.1 | - | 3 | 528 | |||
BC007230.1 | - | 3 | 500 | 2824361 | ||
BC018827.2 | - | 4 | 176 |