Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003648.2 | NP_003639.2 | 29 | 5880 | ||
NM_152879.2 | NP_690618.2 | 30 | 5945 | |||
XM_005246107.3 | XP_005246164.1 | 28 | 5612 | |||
XM_005246108.1 | XP_005246165.1 | 28 | 5766 | |||
XM_005246110.3 | XP_005246167.1 | 19 | 5617 | |||
XM_011512028.2 | XP_011510330.1 | 30 | 7404 | |||
XM_011512029.2 | XP_011510331.1 | 29 | 7274 | |||
XM_011512030.2 | XP_011510332.1 | 30 | 6001 | |||
XM_011512031.2 | XP_011510333.1 | 30 | 5875 | |||
XM_011512032.2 | XP_011510334.1 | 30 | 5859 | |||
XM_011512035.1 | XP_011510337.1 | 28 | 5746 | |||
XM_011512037.2 | XP_011510339.1 | 28 | 5850 | |||
XM_011512040.2 | XP_011510342.1 | 28 | 5782 | |||
XM_017005138.1 | XP_016860627.1 | 30 | 6283 | |||
XM_017005139.1 | XP_016860628.1 | 28 | 5775 | |||
XM_017005140.1 | XP_016860629.1 | 30 | 5877 | |||
GenBank mRNA | AB078966.1 | - | 30 | 5926 | ||
BC006561.2 | - | 4 | 2734 | |||
D63479.3 | - | 30 | 5889 | |||
D73409.1 | - | 29 | 5859 |