Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMUB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076674.1 | NP_001070142.1 | 4 | 1327 | ||
NM_024107.2 | NP_077012.2 | 3 | 1259 | |||
NM_177441.2 | NP_803190.2 | 3 | 1760 | |||
XM_011525201.2 | XP_011523503.1 | 4 | 1335 | |||
XM_011525203.2 | XP_011523505.1 | 5 | 1453 | |||
XM_011525204.2 | XP_011523506.1 | 4 | 1392 | |||
XM_011525205.2 | XP_011523507.1 | 4 | 1853 | |||
XM_011525206.2 | XP_011523508.1 | 5 | 1539 | |||
XM_011525207.2 | XP_011523509.1 | 4 | 1471 | |||
XM_011525208.2 | XP_011523510.1 | 3 | 1364 | |||
XM_011525209.2 | XP_011523511.1 | 3 | 1340 | |||
XM_011525210.2 | XP_011523512.1 | 6 | 1429 | |||
XM_011525211.2 | XP_011523513.1 | 4 | 1186 | |||
XM_017025048.1 | XP_016880537.1 | 4 | 1436 | |||
XM_017025052.1 | XP_016880541.1 | 5 | 1655 | |||
XM_017025054.1 | XP_016880543.1 | 4 | 1639 | |||
XM_017025056.1 | XP_016880545.1 | 4 | 1064 | |||
GenBank mRNA | AF289564.1 | - | 3 | 942 | ||
AF370373.1 | - | 3 | 1738 | |||
AK294198.1 | - | 6 | 1425 | |||
AL833899.1 | - | 3 | 1760 | |||
AY007092.1 | - | 3 | 1182 | |||
AY358780.1 | - | 3 | 1233 | |||
BC001792.1 | - | 3 | 984 | |||
BC004306.1 | - | 3 | 906 | |||
BC063489.1 | - | 3 | 1233 | 5239969 |