Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ERC1 SIRNA Assays ›
Gene Symbol:
ERC1
Gene Name:
ELKS/RAB6-interacting/CAST family member 1
Gene Aliases:
Cast2, ELKS, ERC-1, RAB6IP2
Chromosome Location:
Chr.12: 990010 - 1495933 on Build GRCh38
UniGene ID:
Hs.601216
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
141813
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ERC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001301248.1
NP_001288177.1
15
3059
NM_178039.3
NP_829883.1
15
3090
NM_178040.3
NP_829884.1
16
3174
NR_027946.2
15
3135
NR_027948.2
16
3267
NR_027949.2
12
2286
XM_011520941.2
XP_011519243.1
16
3218
XM_017019052.1
XP_016874541.1
18
3359
XM_017019053.1
XP_016874542.1
18
3293
XM_017019054.1
XP_016874543.1
18
3219
XM_017019055.1
XP_016874544.1
18
3215
XM_017019056.1
XP_016874545.1
18
3156
XM_017019057.1
XP_016874546.1
18
3350
XM_017019058.1
XP_016874547.1
18
3284
XM_017019059.1
XP_016874548.1
17
3347
XM_017019060.1
XP_016874549.1
17
3338
XM_017019061.1
XP_016874550.1
17
3305
XM_017019062.1
XP_016874551.1
17
3296
XM_017019063.1
XP_016874552.1
16
3293
XM_017019064.1
XP_016874553.1
16
3284
XM_017019065.1
XP_016874554.1
17
3266
XM_017019066.1
XP_016874555.1
16
3212
XM_017019067.1
XP_016874556.1
15
3200
XM_017019068.1
XP_016874557.1
16
3284
XM_017019069.1
XP_016874558.1
16
3173
XM_017019070.1
XP_016874559.1
15
3152
XM_017019071.1
XP_016874560.1
15
3080
XM_017019072.1
XP_016874561.1
18
3359
XM_017019074.1
XP_016874563.1
11
1339
XM_017019075.1
XP_016874564.1
10
1283
GenBank mRNA
AB015617.1
-
15
3093
AB029004.2
-
15
2954
AB053468.1
-
16
3208
AB053469.1
-
12
2221
AB053470.1
-
15
3032
AB053471.1
-
16
3084
AK000148.1
-
3
424
AK092201.1
-
5
709
AK294351.1
-
13
2054
BC037377.1
-
5
697
BC132782.1
-
15
3035
40146804
BC132784.1
-
15
3035
40146806
BC144287.1
-
15
3035
BC150248.1
-
15
2975
9007473
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
I-kappaB phosphorylation
multicellular organism development
protein transport
retrograde transport, endosome to Golgi
positive regulation of NF-kappaB transcription factor activity
cell-cell adhesion
protein binding
Rab GTPase binding
PDZ domain binding
leucine zipper domain binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-wqf8l:80/100.66.76.116:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline