Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FADS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178128.5 | NP_835229.3 | 6 | 1636 | ||
GenBank mRNA | AK094411.1 | - | 6 | 1484 | ||
AK293720.1 | - | 3 | 995 | |||
AY203929.1 | - | 6 | 1445 |