Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178499.4 | NP_848594.2 | 9, 10 | 1498 | ||
XM_017018911.1 | XP_016874400.1 | 8, 9 | 1173 | |||
XM_017018912.1 | XP_016874401.1 | 9, 10 | 1281 | |||
XM_017018914.1 | XP_016874403.1 | 5, 6 | 564 | |||
XM_017018915.1 | XP_016874404.1 | 4, 5 | 1529 | |||
XM_017018916.1 | XP_016874405.1 | 4, 5 | 470 | |||
GenBank mRNA | BC040553.1 | - | 9, 10 | 1498 | 5301593 |