Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178844.3 | NP_849172.2 | 5 | 1518 | ||
NR_075083.2 | 5 | 1579 | ||||
XM_017023027.1 | XP_016878516.1 | 5 | 1548 | |||
XM_017023028.1 | XP_016878517.1 | 6 | 1722 | |||
XM_017023029.1 | XP_016878518.1 | 7 | 1803 | |||
XM_017023030.1 | XP_016878519.1 | 7 | 1864 | |||
XM_017023031.1 | XP_016878520.1 | 5 | 1609 | |||
XM_017023032.1 | XP_016878521.1 | 7 | 2565 | |||
XM_017023033.1 | XP_016878522.1 | 7 | 1742 | |||
XM_017023034.1 | XP_016878523.1 | 6 | 1661 | |||
XM_017023035.1 | XP_016878524.1 | 6 | 1690 | |||
XM_017023036.1 | XP_016878525.1 | 5 | 2249 | |||
XM_017023037.1 | XP_016878526.1 | 6 | 1443 | |||
XM_017023038.1 | XP_016878527.1 | 6 | 1416 | |||
XM_017023039.1 | XP_016878528.1 | 5 | 1548 | |||
GenBank mRNA | AK090431.1 | - | 5 | 1575 | ||
AK090476.1 | - | 6 | 1413 | |||
AY601811.1 | - | 5 | 1505 | |||
FJ889357.1 | - | 2 | 1094 |