Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EYA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301012.1 | NP_001287941.1 | 4 | 454 | ||
NM_001301013.1 | NP_001287942.1 | 4 | 454 | |||
NM_004100.4 | NP_004091.3 | 4 | 596 | |||
NM_172103.3 | NP_742101.2 | 4 | 596 | |||
NM_172105.3 | NP_742103.1 | 4 | 596 | |||
XM_005266851.4 | XP_005266908.1 | 4 | 303 | |||
XM_005266853.4 | XP_005266910.1 | 4 | 303 | |||
XM_017010368.1 | XP_016865857.1 | 5 | 823 | |||
XM_017010369.1 | XP_016865858.1 | 4 | 303 | |||
XM_017010370.1 | XP_016865859.1 | 4 | 303 | |||
XM_017010371.1 | XP_016865860.1 | 4 | 303 | |||
XM_017010372.1 | XP_016865861.1 | 4 | 303 | |||
XM_017010373.1 | XP_016865862.1 | 4 | 303 | |||
XM_017010374.1 | XP_016865863.1 | 4 | 602 | |||
XM_017010375.1 | XP_016865864.1 | 4 | 576 | |||
GenBank mRNA | AK295798.1 | - | 4 | 603 | ||
AK299378.1 | - | 4 | 368 | |||
AK301950.1 | - | 5 | 694 | |||
BC041063.1 | - | 4 | 535 | 5458911 | ||
DA760903.1 | - | 4 | 369 | |||
KU178044.1 | - | 3 | 138 | |||
KU178045.1 | - | 3 | 138 | |||
Y17114.1 | - | 4 | 579 |