Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_180989.5 | NP_851320.1 | 3 | 401 | ||
GenBank mRNA | AK126794.1 | - | 3 | 306 | ||
AK289850.1 | - | 3 | 648 | |||
BC033906.1 | - | 3 | 376 | |||
BC052243.1 | - | 3 | 348 | 6083073 | ||
CD110068.1 | - | 3 | 428 |