Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC46A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135919.1 | NP_001129391.1 | 2 | 624 | ||
NM_181785.3 | NP_861450.1 | 2 | 624 | |||
XM_005266361.2 | XP_005266418.1 | 2 | 302 | |||
GenBank mRNA | AF315594.1 | - | 2 | 208 | ||
AK093932.1 | - | 1 | 1052 | |||
BC036662.1 | - | 2 | 586 | 4827143 | ||
BC060850.1 | - | 2 | 445 | 4820254 | ||
BC068556.1 | - | 2 | 452 | 4820792 | ||
BX537565.1 | - | 2 | 435 | |||
DB073710.1 | - | 2 | 480 |