Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181789.2 | NP_861454.2 | 10 | 3460 | ||
XM_011521501.2 | XP_011519803.1 | 10 | 5317 | |||
XM_017022121.1 | XP_016877610.1 | 9 | 3303 | |||
XM_017022122.1 | XP_016877611.1 | 10 | 3491 | |||
XM_017022123.1 | XP_016877612.1 | 10 | 3282 | |||
XM_017022124.1 | XP_016877613.1 | 10 | 3334 | |||
GenBank mRNA | AK095128.1 | - | 1 | 389 | ||
BX538105.1 | - | 10 | 2967 |