Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201407.1 | NP_001188336.1 | 6 | 702 | ||
NM_182531.3 | NP_872337.2 | 5 | 618 | |||
NR_037705.1 | 5 | 575 | ||||
XM_005256288.2 | XP_005256345.1 | 6 | 688 | |||
XM_011522940.2 | XP_011521242.1 | 6 | 655 | |||
XM_017023015.1 | XP_016878504.1 | 5 | 610 | |||
XM_017023016.1 | XP_016878505.1 | 6 | 782 | |||
XM_017023017.1 | XP_016878506.1 | 6 | 681 | |||
GenBank mRNA | AK056437.1 | - | 5 | 611 | ||
BC125192.1 | - | 4 | 323 | 40122711 | ||
BC143524.1 | - | 4 | 292 | |||
DC324714.1 | - | 5 | 566 |