Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182758.3 | NP_877435.3 | 3 | 281 | ||
NR_102334.1 | 3 | 479 | ||||
XM_011521433.2 | XP_011519735.1 | 4 | 478 | |||
XM_011521435.2 | XP_011519737.1 | 3 | 332 | |||
XM_011521436.2 | XP_011519738.1 | 3 | 325 | |||
XM_011521437.2 | XP_011519739.1 | 3 | 325 | |||
XM_017022061.1 | XP_016877550.1 | 3 | 276 | |||
GenBank mRNA | AK096055.1 | - | 4 | 492 | ||
BC101614.1 | - | 2 | 260 | 8069120 | ||
BC101616.1 | - | 2 | 260 | 8069122 | ||
BX537884.1 | - | 3 | 281 | |||
BX648571.1 | - | 3 | 480 |