Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004120.4 | NP_004111.2 | 5 | 781 | ||
GenBank mRNA | AK057968.1 | - | 5 | 694 | ||
AK122866.1 | - | 1 | 2486 | |||
AK226018.1 | - | 5 | 707 | |||
AK314325.1 | - | 5 | 736 | |||
BC022272.1 | - | 5 | 736 | 4688776 | ||
BC073163.1 | - | 5 | 718 | 6151056 | ||
CR457062.1 | - | 4 | 467 | |||
M55543.1 | - | 5 | 623 |