Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC74A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194287.2 | NP_919263.2 | 10, 11 | 1214 | ||
XM_011536479.1 | XP_011534781.1 | 10, 11 | 1582 | |||
XM_011536483.1 | XP_011534785.1 | 10, 11 | 1582 | |||
XM_017021032.1 | XP_016876521.1 | 10, 11 | 1480 | |||
XM_017021033.1 | XP_016876522.1 | 10, 11 | 1561 | |||
XM_017021034.1 | XP_016876523.1 | 10, 11 | 1226 | |||
XM_017021035.1 | XP_016876524.1 | 10, 11 | 1377 | |||
XM_017021036.1 | XP_016876525.1 | 8, 9 | 1450 | |||
XM_017021037.1 | XP_016876526.1 | 10, 11 | 2284 | |||
XM_017021038.1 | XP_016876527.1 | 10, 11 | 1118 | |||
XM_017021039.1 | XP_016876528.1 | 9, 10 | 1040 | |||
XM_017021040.1 | XP_016876529.1 | 10, 11 | 1039 | |||
XM_017021041.1 | XP_016876530.1 | 5, 6 | 538 | |||
GenBank mRNA | AK058106.1 | - | 9, 10 | 1127 | ||
AK125722.1 | - | 11, 12 | 1367 | |||
BC121159.2 | - | 10, 11 | 1062 | 40121413 | ||
BC121160.2 | - | 10, 11 | 1062 | 40121414 |