Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR20A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198441.2 | NP_940843.1 | 3 | 392 | ||
GenBank mRNA | AK097615.1 | - | 3 | 392 | ||
BC120931.1 | - | 3 | 261 | 40114895 | ||
BC120932.1 | - | 3 | 261 | 40114901 |
Gene Symbol: PRR20B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130404.1 | NP_001123876.1 | 3 | 392 |
Gene Symbol: PRR20C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130405.1 | NP_001123877.1 | 3 | 392 |
Gene Symbol: PRR20D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130406.1 | NP_001123878.1 | 3 | 392 |
Gene Symbol: PRR20E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130407.1 | NP_001123879.1 | 3 | 392 |