Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRN1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198443.1 | NP_940845.1 | 2 | 154 | ||
GenBank mRNA | AY358782.1 | - | 2 | 154 | ||
BC100863.2 | - | 2 | 153 | 40002773 | ||
BC100864.1 | - | 2 | 153 | 40002775 | ||
BC100865.1 | - | 2 | 153 | 40002782 | ||
BC100866.2 | - | 2 | 153 | 40002783 |