Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMTNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114974.1 | NP_001108446.1 | 8 | 1883 | ||
NM_198501.2 | NP_940903.2 | 8 | 1535 | |||
XM_005256622.3 | XP_005256679.1 | 7 | 1712 | |||
XM_006721515.2 | XP_006721578.1 | 8 | 2051 | |||
GenBank mRNA | AK124452.1 | - | 8 | 1517 | ||
BC110859.1 | - | 8 | 1883 | 5190568 | ||
DB557882.1 | - | 1 | 39 |