Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFCAB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145053.1 | NP_001138525.1 | 5 | 952 | ||
NM_198529.3 | NP_940931.2 | 5 | 1073 | |||
NR_026738.1 | 5 | 1073 | ||||
XM_011524759.1 | XP_011523061.1 | 5 | 1073 | |||
XM_011524763.2 | XP_011523065.1 | 5 | 1073 | |||
XM_011524765.1 | XP_011523067.1 | 5 | 1073 | |||
XM_011524768.2 | XP_011523070.1 | 5 | 1073 | |||
XM_017024591.1 | XP_016880080.1 | 2 | 152 | |||
XM_017024592.1 | XP_016880081.1 | 5 | 1073 | |||
GenBank mRNA | AK128126.1 | - | 2 | 299 | ||
AK299608.1 | - | 5 | 905 | |||
AK302745.1 | - | 4 | 376 | |||
BC110608.1 | - | 2 | 276 | |||
BC110609.1 | - | 2 | 276 | |||
BC119807.1 | - | 2 | 276 | |||
BC137519.1 | - | 5 | 909 | 9021519 | ||
BX648689.1 | - | 5 | 1011 | |||
DB076773.1 | - | 2 | 299 |