Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182509.3 | NP_872315.2 | 6 | 2328 | ||
NM_199296.2 | NP_954993.1 | 7 | 2674 | |||
XM_011536489.1 | XP_011534791.1 | 7 | 2564 | |||
GenBank mRNA | AJ583024.1 | - | 8 | 2569 | ||
AK056709.1 | - | 8 | 2738 | |||
AK075445.1 | - | 7 | 2667 | |||
BC033140.1 | - | 6 | 2451 | |||
CR749863.1 | - | 7 | 2633 |