Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135217.1 | NP_001128689.1 | 6 | 1074 | ||
NM_006992.3 | NP_008923.1 | 6 | 902 | |||
NM_201650.2 | NP_964013.1 | 6 | 902 | |||
GenBank mRNA | AK223535.1 | - | 6 | 836 | ||
AK292291.1 | - | 6 | 837 | |||
AK292367.1 | - | 6 | 902 | |||
BC029858.1 | - | 6 | 875 | 5170643 | ||
U72508.1 | - | 6 | 794 |