Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000264.3 | NP_000255.2 | 24 | 4855 | ||
NM_001083602.1 | NP_001077071.1 | 24 | 4963 | |||
NM_001083603.1 | NP_001077072.1 | 24 | 4809 | |||
NM_001083604.1 | NP_001077073.1 | 24 | 4573 | |||
NM_001083605.1 | NP_001077074.1 | 24 | 4556 | |||
NM_001083606.1 | NP_001077075.1 | 24 | 4655 | |||
NM_001083607.1 | NP_001077076.1 | 24 | 4729 | |||
GenBank mRNA | AB209495.1 | - | 21 | 7973 | ||
AK124593.1 | - | 3 | 1730 | |||
U43148.1 | - | 24 | 4655 |