Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000328.2 | NP_000319.1 | 14 | 1889 | ||
NM_001034853.1 | NP_001030025.1 | 14 | 1889 | |||
XM_011543940.2 | XP_011542242.1 | 14 | 1885 | |||
XM_017029710.1 | XP_016885199.1 | 14 | 1918 | |||
XM_017029711.1 | XP_016885200.1 | 14 | 1915 | |||
GenBank mRNA | AJ238395.1 | - | 14 | 1776 | ||
AK291832.1 | - | 14 | 1896 | |||
U57629.1 | - | 14 | 1780 | |||
X97668.1 | - | 14 | 1721 |