Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSEN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000447.2 | NP_000438.2 | 5 | 674 | ||
NM_012486.2 | NP_036618.2 | 5 | 674 | |||
XM_005273199.3 | XP_005273256.1 | 4 | 487 | |||
XM_017001835.1 | XP_016857324.1 | 4 | 995 | |||
XM_017001836.1 | XP_016857325.1 | 4 | 487 | |||
GenBank mRNA | AF416718.1 | - | 3 | 315 | ||
AK292299.1 | - | 6 | 626 | |||
AK301143.1 | - | 4 | 423 | |||
BC006365.2 | - | 6 | 868 | 4131408 | ||
BT006984.1 | - | 3 | 247 | |||
DA915823.1 | - | 5 | 627 | |||
L43964.1 | - | 4 | 614 | |||
L44577.1 | - | 5 | 612 | |||
U34349.1 | - | 3 | 338 |