Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000542.3 | NP_000533.3 | 4 | 345 | ||
NM_198843.2 | NP_942140.2 | 4 | 345 | |||
XM_005264487.2 | XP_005264544.1 | 3 | 260 | |||
XM_005264488.3 | XP_005264545.2 | 3 | 225 | |||
XM_005264490.4 | XP_005264547.2 | 3 | 225 | |||
XM_017004702.1 | XP_016860191.1 | 3 | 260 | |||
XM_017004703.1 | XP_016860192.1 | 3 | 225 | |||
GenBank mRNA | AK314947.1 | - | 3 | 225 | ||
AL548140.3 | - | 4 | 345 | |||
BC032785.1 | - | 3 | 212 | 5179408 | ||
BI821350.1 | - | 215 | ||||
DQ317589.1 | - | 3 | 198 | |||
J02761.1 | - | 3 | 222 |