Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000655.4 | NP_000646.2 | 3 | 319 | ||
NR_029467.1 | 1 | 83 | ||||
GenBank mRNA | AB583235.1 | - | 2 | 115 | ||
AJ246000.1 | - | 3 | 190 | |||
AK225713.1 | - | 3 | 230 | |||
AK298705.1 | - | 1 | 78 | |||
AK312673.1 | - | 3 | 230 | |||
BC020758.1 | - | 3 | 216 | 4717456 | ||
DB271854.1 | - | 3 | 316 | |||
DB482055.1 | - | 3 | 313 | |||
DC378123.1 | - | 1 | 83 | |||
M25280.1 | - | 3 | 230 | |||
X16150.1 | - | 3 | 206 | |||
X17519.1 | - | 3 | 203 |