Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000655.4 | NP_000646.2 | 9 | 2429 | ||
NR_029467.1 | 7 | 2193 | ||||
GenBank mRNA | BP393094.1 | - | 1 | 487 | ||
X16150.1 | - | 9 | 2310 |