Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000661.4 | NP_000652.2 | 3 | 134 | ||
NM_001024921.3 | NP_001020092.1 | 3 | 292 | |||
GenBank mRNA | AB062431.1 | - | 1 | 81 | ||
AK130966.1 | - | 1 | 97 | |||
AK297178.1 | - | 3 | 117 | |||
AK303908.1 | - | 1 | 97 | |||
AK312241.1 | - | 1 | 101 | |||
AL039355.2 | - | 3 | 267 | |||
AL832047.1 | - | 2 | 189 | |||
AY320412.1 | - | 3 | 117 | |||
AY376242.1 | - | 2 | 251 | |||
AY597415.1 | - | 1 | 108 | |||
BC000483.2 | - | 1 | 86 | 2964733 | ||
BC004156.2 | - | 1 | 80 | 2961026 | ||
BC004206.2 | - | 1 | 80 | 2961026 | ||
BC007967.2 | - | 1 | 136 | 4304670 | ||
BC012149.1 | - | 1 | 136 | 4554838 | ||
BC031906.1 | - | 2 | 81 | 4996986 | ||
BC066318.1 | - | 1 | 101 | 5277491 | ||
BC070214.1 | - | 1 | 140 | 6605055 | ||
BC100285.1 | - | 2 | 241 | 6022858 | ||
BG829769.1 | - | 2 | 82 | |||
BM841336.1 | - | 3 | 134 | |||
D14531.1 | - | 2 | 81 | |||
D16961.1 | - | 1 | 54 | |||
U09953.1 | - | 3 | 104 | |||
U21138.1 | - | 1 | 81 |