Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000661.4 | NP_000652.2 | 5 | 345 | ||
NM_001024921.3 | NP_001020092.1 | 5 | 503 | |||
GenBank mRNA | AB062431.1 | - | 1 | 292 | ||
AK130966.1 | - | 1 | 308 | |||
AK297178.1 | - | 5 | 328 | |||
AK303908.1 | - | 1 | 308 | |||
AK312241.1 | - | 1 | 312 | |||
AL039355.2 | - | 5 | 478 | |||
AL832047.1 | - | 3 | 1918 | |||
AY320412.1 | - | 5 | 328 | |||
AY376242.1 | - | 4 | 462 | |||
AY597415.1 | - | 1 | 319 | |||
BC000483.2 | - | 1 | 297 | 2964733 | ||
BC004156.2 | - | 1 | 291 | 2961026 | ||
BC004206.2 | - | 1 | 291 | 2961026 | ||
BC007967.2 | - | 1 | 347 | 4304670 | ||
BC012149.1 | - | 1 | 347 | 4554838 | ||
BC031906.1 | - | 4 | 292 | 4996986 | ||
BC066318.1 | - | 1 | 312 | 5277491 | ||
BC070214.1 | - | 1 | 351 | 6605055 | ||
BC100285.1 | - | 4 | 452 | 6022858 | ||
BG829769.1 | - | 4 | 293 | |||
BM841336.1 | - | 5 | 345 | |||
D14531.1 | - | 4 | 292 | |||
U09953.1 | - | 5 | 315 | |||
U21138.1 | - | 1 | 292 |