Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000966.5 | NP_000957.1 | 5, 6 | 975 | ||
NM_001042728.2 | NP_001036193.1 | 3, 4 | 763 | |||
NM_001243730.1 | NP_001230659.1 | 4, 5 | 649 | |||
NM_001243731.1 | NP_001230660.1 | 3, 4 | 500 | |||
NM_001243732.1 | NP_001230661.1 | 2, 3 | 614 | |||
GenBank mRNA | AB307689.1 | - | 3, 4 | 483 | ||
AK290588.1 | - | 5, 6 | 971 | |||
AK297023.1 | - | 4, 5 | 567 | |||
AK297110.1 | - | 3, 4 | 476 | |||
AK297277.1 | - | 2, 3 | 614 | |||
AK297801.1 | - | 3, 4 | 609 | |||
AK301057.1 | - | 3, 4 | 758 | |||
AK316259.1 | - | 3, 4 | 487 | |||
BC093727.1 | - | 3, 4 | 507 | 7939572 | ||
BC093729.1 | - | 3, 4 | 507 | 7939574 | ||
HQ692828.1 | - | 3, 4 | 483 | |||
HQ692829.1 | - | 3, 4 | 450 | |||
L12060.1 | - | 4, 5 | 591 | |||
M24857.1 | - | 5, 6 | 897 | |||
M38258.1 | - | 3, 4 | 629 | |||
M57707.1 | - | 4, 5 | 682 |