Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000970.4 | NP_000961.2 | 4 | 443 | ||
NM_001024662.2 | NP_001019833.1 | 4 | 619 | |||
NM_001320137.1 | NP_001307066.1 | 4 | 624 | |||
NM_001320138.1 | NP_001307067.1 | 4 | 658 | |||
NM_001320139.1 | NP_001307068.1 | 4 | 505 | |||
NM_001320140.1 | NP_001307069.1 | 4 | 667 | |||
NM_001320141.1 | NP_001307070.1 | 6 | 732 | |||
NM_001320142.1 | NP_001307071.1 | 4 | 379 | |||
XM_017019781.1 | XP_016875270.1 | 6 | 1767 | |||
XM_017019782.1 | XP_016875271.1 | 6 | 928 | |||
GenBank mRNA | AF261087.1 | - | 1 | 410 | ||
AK299470.1 | - | 5 | 373 | |||
AK308469.1 | - | 4 | 586 | |||
AK311863.1 | - | 4 | 407 | |||
BC004138.2 | - | 1 | 419 | 2823733 | ||
BC013863.1 | - | 1 | 393 | |||
BC020679.1 | - | 1 | 407 | 4616668 | ||
BC022444.1 | - | 4 | 574 | 4286423 | ||
BC031009.1 | - | 1 | 407 | 4333761 | ||
BC032299.1 | - | 1 | 404 | 4994660 | ||
BC071912.1 | - | 1 | 396 | 6293404 | ||
BC104824.1 | - | 4 | 419 | 8143827 | ||
BC104826.1 | - | 4 | 419 | 8143829 | ||
BG030827.1 | - | 1 | 169 | |||
BP285025.1 | - | 4 | 443 | |||
CB140996.1 | - | 4 | 572 | |||
CD049618.1 | - | 4 | 589 | |||
D17554.1 | - | 4 | 409 | |||
HY016219.1 | - | 4 | 453 | |||
S71022.1 | - | 3 | 253 | |||
X69391.1 | - | 4 | 414 |