Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000983.3 | NP_000974.1 | 4 | 726 | ||
GenBank mRNA | AF113701.1 | - | 1 | 189 | ||
AK097075.1 | - | 4 | 685 | |||
AL110153.1 | - | 1 | 126 | |||
BC035566.1 | - | 4 | 726 | 5109124 | ||
BC066314.1 | - | 4 | 702 | 5303658 |