Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000985.4 | NP_000976.1 | 5 | 619 | ||
NM_001035006.2 | NP_001030178.1 | 5 | 370 | |||
NM_001199340.1 | NP_001186269.1 | 6 | 468 | |||
NM_001199341.1 | NP_001186270.1 | 5 | 469 | |||
NM_001199342.1 | NP_001186271.1 | 6 | 567 | |||
NM_001199343.1 | NP_001186272.1 | 6 | 550 | |||
NM_001199344.1 | NP_001186273.1 | 5 | 452 | |||
NM_001199345.1 | NP_001186274.1 | 4 | 329 | |||
GenBank mRNA | AK304659.1 | - | 4 | 243 | ||
AK311828.1 | - | 5 | 284 | |||
BC000502.2 | - | 4 | 522 | 2821464 | ||
BC017831.1 | - | 1 | 288 | 4251433 | ||
BC066323.1 | - | 1 | 284 | 4797583 | ||
BC066324.1 | - | 1 | 288 | 4799029 | ||
BC106031.1 | - | 1 | 255 | 4540700 | ||
BG758380.1 | - | 6 | 486 | |||
BG827384.1 | - | 5 | 370 | |||
BI086293.1 | - | 5 | 341 | |||
BQ877039.1 | - | 6 | 508 | |||
BQ882430.1 | - | 5 | 435 | |||
CD386294.1 | - | 5 | 389 | |||
CD387752.1 | - | 5 | 474 | |||
X53777.1 | - | 4 | 387 |
Gene Symbol: RPL17-C18orf32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199355.1 | NP_001186284.1 | 4 | 262 | ||
NM_001199356.1 | NP_001186285.1 | 4 | 329 | |||
GenBank mRNA | BI461805.1 | - | 4 | 248 | ||
BX393840.2 | - | 4 | 264 |