Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000991.4 | NP_000982.2 | 5 | 725 | ||
NM_001136134.1 | NP_001129606.1 | 4 | 919 | |||
GenBank mRNA | AB209627.1 | - | 2 | 2138 | ||
AK293736.1 | - | 4 | 917 | |||
BC011582.1 | - | 4 | 704 | 4561118 |