Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000991.4 | NP_000982.2 | 5 | 3211 | ||
NM_001136134.1 | NP_001129606.1 | 4 | 3405 | |||
GenBank mRNA | AB209627.1 | - | 2 | 4620 | ||
AK025866.1 | - | 1 | 1007 | |||
AK125297.1 | - | 1 | 1956 | |||
BC011582.1 | - | 4 | 3186 | 4561118 |