Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000994.3 | NP_000985.1 | 2 | 171 | ||
NM_001007073.1 | NP_001007074.1 | 2 | 270 | |||
NM_001007074.1 | NP_001007075.1 | 1 | 290 | |||
GenBank mRNA | AK311909.1 | - | 2 | 128 | ||
BC011514.1 | - | 1 | 97 | 2900089 | ||
BC070209.1 | - | 1 | 103 | 6738371 | ||
BF960792.1 | - | 1 | 301 | |||
BQ068496.1 | - | 208 | ||||
CF124158.1 | - | 2 | 208 | |||
X03342.1 | - | 2 | 111 |
Gene Symbol: RPL32P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003111.2 | 2 | 514 | |||
GenBank mRNA | AK093477.1 | - | 2 | 494 | ||
BC015389.1 | - | 1 | 78 | |||
BC053996.1 | - | 2 | 480 | |||
DA215350.1 | - | 2 | 502 |