Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001020.5 | NP_001011.1 | 5 | 575 | ||
NM_001321111.1 | NP_001308040.1 | 5 | 524 | |||
XM_005259137.2 | XP_005259194.1 | 4 | 670 | |||
GenBank mRNA | AK055869.1 | - | 4 | 2348 | ||
BC004324.2 | - | 5 | 501 | 3628799 | ||
BC007977.2 | - | 5 | 486 | 4310307 | ||
BC024970.1 | - | 1 | 471 | |||
BC071674.1 | - | 4 | 599 | 5394209 | ||
BC095429.1 | - | 5 | 445 | |||
BQ421722.1 | - | 5 | 533 | |||
M60854.1 | - | 5 | 487 |