Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POU5F1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | GU480897.1 | - | 1 | 865 |
Gene Symbol: POU5F1P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | GU480856.1 | - | 1 | 868 | ||
HQ122904.1 | - | 1 | 868 |