Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002705.4 | NP_002696.3 | 15 | 1876 | ||
XM_006720902.3 | XP_006720965.1 | 15 | 1840 | |||
XM_017023374.1 | XP_016878863.1 | 15 | 1888 | |||
XM_017023375.1 | XP_016878864.1 | 15 | 1900 | |||
GenBank mRNA | AB011140.1 | - | 6 | 798 | ||
AF001691.1 | - | 15 | 1876 | |||
AF013717.1 | - | 15 | 1797 | |||
BC114620.1 | - | 15 | 1836 | 40070172 |