Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002711.3 | NP_002702.2 | 1 | 308 | ||
GenBank mRNA | BC056666.1 | - | 1 | 311 | ||
BC070313.1 | - | 1 | 290 | |||
BC126451.1 | - | 1 | 346 | 8992167 | ||
BC126453.1 | - | 1 | 346 | 8992169 | ||
BF576879.1 | - | 291 | ||||
X78578.1 | - | 1 | 275 |