Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303503.1 | NP_001290432.1 | 6 | 538 | ||
NM_001303504.1 | NP_001290433.1 | 5 | 430 | |||
NM_001303506.1 | NP_001290435.1 | 6 | 591 | |||
NM_001303507.1 | NP_001290436.1 | 5 | 480 | |||
NM_002720.2 | NP_002711.1 | 6 | 532 | |||
XM_006721061.3 | XP_006721124.1 | 5 | 417 | |||
XM_017023403.1 | XP_016878892.1 | 6 | 508 | |||
XM_017023404.1 | XP_016878893.1 | 5 | 444 | |||
GenBank mRNA | AF097996.1 | - | 6 | 454 | ||
AL519181.3 | - | 5 | 434 | |||
BC001416.2 | - | 6 | 485 | 3140324 | ||
BG706310.1 | - | 5 | 483 | |||
BG741669.1 | - | 6 | 563 | |||
BQ220878.1 | - | 5 | 340 | |||
BT019746.1 | - | 5 | 350 | |||
X70218.1 | - | 6 | 485 |