Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321053.1 | NP_001307982.1 | 2, 3 | 432 | ||
NM_002727.3 | NP_002718.2 | 1, 2 | 217 | |||
XM_017016392.1 | XP_016871881.1 | 2, 3 | 845 | |||
GenBank mRNA | BG548665.1 | - | 1, 2 | 97 | ||
BP363056.1 | - | 1, 2 | 174 |