Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKAR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002736.2 | NP_002727.2 | 11 | 1752 | ||
XM_011516398.1 | XP_011514700.1 | 10 | 1673 | |||
GenBank mRNA | AK128639.1 | - | 12 | 1562 | ||
BC075800.1 | - | 11 | 1791 | 30331962 | ||
CA405423.1 | - | 3 | 629 | |||
M31158.1 | - | 11 | 1743 |