Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002752.4 | NP_002743.3 | 12 | 1573 | ||
NM_139068.2 | NP_620707.1 | 12 | 1578 | |||
NM_139069.2 | NP_620708.1 | 12 | 1578 | |||
NM_139070.2 | NP_620709.1 | 12 | 1573 | |||
XM_005265940.4 | XP_005265997.1 | 13 | 1706 | |||
XM_006714891.2 | XP_006714954.1 | 13 | 1631 | |||
XM_017009638.1 | XP_016865127.1 | 13 | 1542 | |||
XM_017009639.1 | XP_016865128.1 | 14 | 1799 | |||
XM_017009640.1 | XP_016865129.1 | 13 | 1690 | |||
XM_017009641.1 | XP_016865130.1 | 13 | 1645 | |||
GenBank mRNA | AK289638.1 | - | 12 | 1469 | ||
BC032539.1 | - | 11 | 1363 | 5528624 | ||
L31951.1 | - | 11 | 1360 | |||
U09759.1 | - | 11 | 1453 | |||
U34821.1 | - | 11 | 1351 | |||
U35002.1 | - | 11 | 1329 | |||
U35003.1 | - | 11 | 1336 |