Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002754.4 | NP_002745.1 | 2 | 282 | ||
NR_072996.1 | 2 | 282 | ||||
GenBank mRNA | AB209586.1 | - | 1 | 1240 | ||
AB451221.1 | - | 3 | 183 | |||
AB451344.1 | - | 3 | 183 | |||
AF015256.1 | - | 2 | 243 | |||
AF092535.1 | - | 2 | 204 | |||
AF100546.1 | - | 2 | 185 | |||
AK222922.1 | - | 2 | 174 | |||
AK308018.1 | - | 2 | 263 | |||
BC000433.2 | - | 2 | 224 | 2819932 | ||
BC001641.2 | - | 2 | 224 | 2819932 | ||
BC004428.1 | - | 2 | 224 | 2819932 | ||
BC085196.1 | - | 2 | 252 | 6107307 | ||
BM921729.1 | - | 2 | 281 | |||
BT007221.1 | - | 2 | 183 | |||
CR536490.1 | - | 2 | 183 | |||
CR541687.1 | - | 2 | 183 | |||
DB473379.1 | - | 2 | 246 | |||
DR006449.1 | - | 2 | 286 | |||
KU178281.1 | - | 2 | 183 | |||
KU178282.1 | - | 2 | 183 | |||
U93232.1 | - | 2 | 190 | |||
Y10488.1 | - | 2 | 205 |